Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Sucrose, crystals, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: alpha-D-GlucopyranosideSucroseD-(+)-SaccharoseD(+)-Saccharosebeta-D-Fructofuranosyl alpha-D-glucopyranosideSugar

Management of Change (MOC) category = N

MACR8360-06 MACR8360-20 MACR8360-06 MACR8360-24 MACR8360-20 MACR8360-24
MK836024EA 1380.59 USD InStock
MK836024 CAMK836020 MK836020 CAMK836006 MK836006 CAMK836024
D-(+)-Sucrose, crystals, AR® ACS, Macron Fine Chemicals™
D(+)-Sucrose


Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 12,09051
https://us-legacy.vwr.com/stibo/web/std.lang.all/95/86/2299586.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Solubility Passes Test
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤0.005 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Iron (Fe) ≤5 ppm
ACS - Titrable Acid (meq/g) ≤0.0008
ACS - Invert Sugar ≤0.05 %
ACS - Loss on Drying at 105°C ≤0.03 %
ACS - Residue after Ignition ≤0.01 %
ACS - Sulfate and Sulfite (as SO₄) ≤0.005 %
ACS - Specific Rotation [Å]²⁵^D (+) 66.3 - 66.8 Degree

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty