Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xylene (mixture of isomers) ≥98.5% (by GC), AR® ACS for histology, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Dimethylbenzene

Management of Change (MOC) category = N

8668-06 MACR8668-22 MACR8668-02 MACR8668-16 MACR8668-06 MACR8668-26 MACR8668-16 MACR8668-02 MACR8668-22 MACR8668-26
MK866822EA 435.67 USD InStock
MK866822 CAMK866822 CAMK866802 CAMK866826 CAMK866816 CAMK866806 MK866806 MK866816 MK866826 MK866802
Xylene (mixture of isomers) ≥98.5% (by GC), AR® ACS for histology, Macron Fine Chemicals™
Xylene (mixture of isomers)
ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  


Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –34 °C
Density: 0.86…0.88 g/cm³ (20 °C)
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://us-legacy.vwr.com/stibo/web/std.lang.all/44/47/704447.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (m,p,o-xylenes and ethylbenzene) (by GC) ≥ 98.5 %
ACS - Color (APHA) ≤ 10
ACS - Ethylbenzene (C₆H₅C₂H₅) ≤ 25 %
Particulate Contamination Passes Test
ACS - Residue after Evaporation ≤ 0.002 %
ACS - Substances Darkened by H₂SO₄ Passes Test
Sulfur Compounds (as S) ≤ 0.0005 %
ACS - Water (H₂O) ≤ 0.02 %
Appearance (clear, colorless liquid)
Date Received: / /
Date Opened: / /

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty