Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc oxide 99.0-100.5% (excluding ignited substances) USP, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Zinci oxidumZinc (II) oxide

Management of Change (MOC) category = R

MACR8824-05 MACR8824-24 MACR8824-30 MACR8824-24 MACR8824-05 MACR8824-30 MACR8824-30 MACR8824-24
JT8824-24CS 2110.78 USD InStock
CAMK882405 CAMK8824-24 JT8824-24 MK8824-30 MK882405 MK8824-24 JT8824-30 CAMK8824-30
Zinc oxide 99.0-100.5% (excluding ignited substances) USP, Macron Fine Chemicals™
Zinc oxide


Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.61 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200
Order Now
Specification Test Results
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (ZnO) (ignited basis) 98.0 - 102.0 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Alkalinity Passes Test
USP - Loss on Ignition ≤1.0 %
USP - Carbonate and Color of Solution Passes Test
USP - Arsenic (As) ≤6 ppm
USP - Lead (Pb) Passes Test
USP - Iron and Other Heavy Metals Passes Test
Solubility in Dilute Sulfuric Acid Passes Test
Lead (Pb) ≤0.0010 %
Cadmium (Cd) ≤0.0015 %
Iron (Fe) ≤0.0020 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty