Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc sulfate heptahydrate, granular USP, FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = R

MACR8872-20 MACR8872-24 MACR8872-07 MACR8872-20 MACR8872-24 MACR8872-07
MK887224EA 8817.91 USD InStock
CAMK887220 MK887224 CAMK887207 MK887220 MK887207 CAMK887224
Zinc sulfate heptahydrate, granular USP, FCC, Macron Fine Chemicals™
Zinc sulphate heptahydrate


Formula: ZnSO₄·7H₂O
MW: 287.54 g/mol
Melting Pt: 100 °C
Density: 1.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 12,10293
Order Now
Specification Test Results
Meets U.S.P Requirements
Food GMP Manufactured Product
GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
Assay ((as ZnSO₄)) 55.6 - 61.0 %
Assay (ZnSO₄·7H₂O) 99.0 - 108.7 %
Identification A Passes Test
Identification B Passes Test
Appearance Passes Test
Acidity Passes Test
FCC - Alkalies and Alkaline Earths ≤0.5 %
USP - Alkalies and Alkaline Earths ≤0.9 %
Arsenic (As) ≤3 ppm
FCC - Iron (Fe) ≤0.003 %
FCC - Lead (Pb)(FCC) ≤4 mg/kg
USP - Lead (Pb) ≤0.002 %
FCC - Cadmium (Cd) ≤2 mg/kg
FCC - Mercury (Hg) ≤5 ppm
FCC - Selenium (Se) ≤0.003 %
Solubility Passes Test
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty