Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc sulfate heptahydrate 99.0-103.0%, granular, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = N

MACR8880-12 8880-06 MACR8880-06
MK888006CS 2559.46 USD InStock
MK888006 MK8880-12 CAMK888006
Zinc sulfate heptahydrate 99.0-103.0%, granular, AR® ACS, Macron Fine Chemicals™
Zinc sulphate heptahydrate


Formula: ZnSO₄·7H₂O
MW: 287.54 g/mol
Melting Pt: 100 °C
Density: 1.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 12,10293
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (ZnSO₄·7H₂O) 99.0 - 103.0 %
ACS - Insoluble Matter ≤0.01 %
ACS - Chloride (Cl) ≤0.0005 %
ACS - Nitrate (NO₃) ≤0.002 %
ACS - Ammonium (NH₄) ≤0.001 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Iron (Fe) ≤0.001 %
ACS - Lead (Pb) ≤0.003 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Manganese (Mn) ≤0.0003 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.05 %
Solubility Passes Test
ACS - pH of 5% Solution at 25°C 4.4 - 6.0
Appearance (white to off-white granules)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty