Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol ≥99.5% (by GC) NF, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

Management of Change (MOC) category = RL

MACR8888-R MACR8888-R
MK8888-REA 4 USD InStock
CAMK8888-R MK8888-R
Methanol ≥99.5% (by GC) NF, Macron Fine Chemicals™
Methanol
Tested to NF Specifications (for Upsher-Smith)

No Class 1 or Class 2 Residual Solvents are present in this product Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%


Formula: CH₃OH
MW: 32.04 g/mol
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
NF - Assay (CH₃OH) (by GC) ≥99.5 %
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Acetone, Aldehydes (as (CH₃)₂CO) ≤0.003 %
NF - Acidity Passes Test
NF - Alkalinity (as NH₃) ≤3 ppm
NF - Nonvolatile Residue ≤0.001 %
NF - Readily Carbonizable Substances Passes Test
NF - Readily Oxidizable Substances Passes Test
NF - Water (H₂O) ≤0.1 %
No Class 1 or Class 2 Residual Solvents are present in this product Only
Class 3 Solvents are likely to be present. Residual concentration of all
Class 3 Solvents is <0.5%

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty