Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutylammonium hydroxide 40% in water, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = N

MACRE224-80 MACRE224-80
MKE22480EA 267.07 USD InStock
MKE22480 CAMKE22480
Tetrabutylammonium hydroxide 40% in water, Macron Fine Chemicals™
Tetrabutylammonium hydroxide


Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8,II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Warm product to 30°C and shake prior to use.
Assay ((CH₃CH₂CH₂CH₂)₄NOH) 38 - 42 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty