Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodine 0.01 N (N/100) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Management of Change (MOC) category = N

MACRH175-05 MACRH175-07 MACRH175-07
MKH17507CS 445.45 USD InStock
CAMKH17507 MKH17507 MKH17505
Iodine 0.01 N (N/100) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Iodine
Contains 0.1–0.4% iodine and Max. 1% potassium iodide.


Formula: I₂
MW: 253.81 g/mol
MDL Number: MFCD00011355
CAS Number: 7553-56-2
ADR: 8,III
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Normality 0.0095 - 0.0105
Appearance (transparent, reddish-brown solution)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty