Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead acetate cotton, StandARd®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Lead di-acetateLead (II) acetate on cotton

Management of Change (MOC) category = N

MACRH220-03 MACRH220-03
MKH22003EA 563.33 USD InStock
MKH22003 CAMKH22003
Lead acetate cotton, StandARd®, Macron Fine Chemicals™
Lead acetate cotton
In cotton balls. For arsenic.


Formula: (H₃CCOO)₂Pb
MW: 325.29 g/mol
Melting Pt: 175…180 °C
Density: ∼2.22 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 6080-56-4
EINECS: 208-908-0
UN: 1616
ADR: 6.1,III
Merck Index: 13,05442
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Identification Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty