Would you like to log in again?
Macron Fine Chemicals™ StandARd®, Platinum Cobalt Color Standard (APHA 500), StandARd®, Macron Fine Chemicals™
Supplier: Avantor
MACRH296-01 MACRH296-03
MKH29601CS 5064.18 USD InStock
MKH29601 MKH29603
Macron Fine Chemicals™ StandARd®, Platinum Cobalt Color Standard (APHA 500), StandARd®, Macron Fine Chemicals™
Standards Color Reference and Spectroscopy Standards
Corrosive
APHA 500. Contains 0.13% chloroplatinic acid, 0.11% cobalt (II) chloride, and 4.6% hydrogen chloride in water.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty