Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenol solution, StandARd®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Carbolic acidHydroxybenzenePhenylalcohol

Management of Change (MOC) category = N

MACRH302-01 MACRH302-01
MKH30201CS 347.82 USD InStock
CAMKH30201 MKH30201
Phenol solution, StandARd®, Macron Fine Chemicals™
Phenol


Formula: C₆H₅OH
MW: 94.11 g/mol
MDL Number: MFCD00002143
CAS Number: 108-95-2
UN: 1671
ADR: 6.1,II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Concentration (0.8-1.2 mg/mL) Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty