Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol ≥99.9% (by GC), UltimAR® for liquid chromatography, for organic residue analysis, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

Management of Change (MOC) category = NI

MACRH488-10 MACRH488-06 MACRH488-06 MACRH488-68 MACRH488-10
MKH48806CS 685.36 USD InStock
MKH48806 CAMKH48806 MKH48810 CAMKH48810 JTH488-68
Methanol ≥99.9% (by GC), UltimAR® for liquid chromatography, for organic residue analysis, for UV spectrophotometry, Macron Fine Chemicals™
Methanol
UltimAR—Solvents that meet ACS specifications for liquid chromatography, UV-spectrophotometry, and general purpose use. They are also tested by GC/ECD and GC/FID for use in extraction/concentration sample clean-up protocols for trace-level organic impurities. 


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
ACS - Assay (CH₃OH) (by GC, corrected for water) ≥99.9 %
Carbonyl Compounds - Acetone ≤0.001 %
Carbonyl Compounds - Acetaldehyde ≤0.001 %
Carbonyl Compounds - Formaldehyde ≤0.001 %
Color (APHA) ≤5
Residue after Evaporation ≤0.00010 %
Solubility in H₂O Passes Test
Substances Darkened by H₂SO₄ Passes Test
Substances Reducing Permanganate Passes Test
Titrable Acid (meq/g) ≤0.0003
Titrable Base (meq/g) ≤0.0002
Water (H₂O)(by Karl Fischer titrn) ≤0.05 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 205 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤0.80
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.25
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤0.12
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 - 280 nm ≤0.005
Liquid Chromatography Suitability Absorbance Passes Test
HPLC Gradient Elution (No Peak Greater than 25% of Full Scale)(0.005 au) Passes Test
ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L) ≤5
Sum of the Peaks (ng/L) ≤10
FID Sensitive Impurities (as 2-Octano l) Single Peak (µg/L) ≤5
Sum of the Peaks (µg/L) ≤10
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty