Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 36.5 - 38.0%, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Chlorohydric acidHydrochloric acid gasHydrochlorideHydrogen chloride gasMuriatic acid

Management of Change (MOC) category = N

MACRH613-27 MACRH613-05 MACRH613-45 MACRH613-14 MACRH613-05 MACRH613-45 MACRH613-16 MACRH613-46 MACRH613-46 MACRH613-16 MACRH613-14 MACRH613-27 MACRH613-14 MACRH613-69
MKH61316CS 640.4 USD InStock
CAMKH61327 CAMKH61316 MKH61316 MKH61327 JTH613-14 CAMKH613-05 MKH613-05 CAMKH613-14 CAMKH613-45 MKH613-14 MKH613-69 CAMKH61346 MKH613-45 MKH613-46
Hydrochloric acid 36.5 - 38.0%, AR® ACS, Macron Fine Chemicals™
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HCl) 36.5 - 38.0 %
Appearance Passes Test
ACS - Color (APHA) ≤ 10
ACS - Ammonium (NH₄) ≤ 3 ppm
ACS - Arsenic (As) ≤ 0.005 ppm
ACS - Bromide (Br) ≤ 0.005 %
ACS - Extractable Organic Substances ≤ 5 ppm
ACS - Free Chlorine (as Cl) ≤ 1 ppm
ACS - Heavy Metals (as Pb) ≤ 0.1 ppm
ACS - Residue after Ignition ≤ 4 ppm
ACS - Sulfate (SO₄) ≤ 0.5 ppm
ACS - Sulfite (SO₃) ≤ 0.8 ppm
Trace Impurities - Aluminum (Al) ≤ 0.300 ppm
Trace Impurities - Boron (B) ≤ 0.100 ppm
Trace Impurities - Chromium (Cr) ≤ 0.200 ppm
Trace Impurities - Copper (Cu) ≤ 0.10 ppm
Trace Impurities - Gold (Au) ≤ 0.100 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Trace Impurities - Lead (Pb) ≤ 0.100 ppm
Trace Impurities - Magnesium (Mg) ≤ 0.300 ppm
Trace Impurities - Manganese (Mn) ≤ 0.300 ppm
Trace Impurities - Nickel (Ni) ≤ 0.1 ppm
Trace Impurities - Potassium (K) ≤ 0.300 ppm
Trace Impurities - Tin (Sn) ≤ 0.300 ppm
Trace Impurities - Titanium (Ti) ≤ 0.300 ppm
Trace Impurities - Zinc (Zn) ≤ 0.100 ppm

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty