Would you like to log in again?
MABPAC SCX-10 COLUMN 4X250MM
Supplier: Thermo Scientific
Mabpac* SCX-10 LC Column 4x250mm^ Mabpac* SCX-10 LC Column, Material: Non-porous polymer, Technique: Monoclonal antibody/Cation exchange, Flow rate: 0.2-2ml/min, Pressure: 3000psi, Temperature: 60deg C, pH: 2-12, Phase: Sulfonate, Solvent: 50% Acetonitrile, Particle size: 10um, Dimension: 4x250mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MABPAC SCX-10 COLUMN 4X250MM MP10042-824 074625 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty