Would you like to log in again?
ReLeSR™, Stemcell Technologies
Supplier: Stemcell Technologies
100-0483
MSPP-05872EA 90.62 USD InStock
MSPP-05872
ReLeSR™, Stemcell Technologies
Cell Separation Media
Enzyme-free human ES and iPS cell selection and passaging reagent.
  • Simple passaging protocol
  • Eliminates the need for manual removal (selection) of differentiated cells
  • No manual scraping to generate cell aggregates
  • Compatible with passaging in flasks and large culture vessels
  • Chemically defined, enzyme-free, and gentle on cells
  • High expansion of human ES/iPS cells after passaging
ReLeSR™ is an enzyme-free reagent for dissociation and passaging of human pluripotent stem cells (hPSCs) as aggregates without manual selection or scraping. Passaging hPSCs with ReLeSR™ easily generates optimally-sized aggregates, while eliminating the hassle and variability associated with manual manipulation. By eliminating the need for scraping, ReLeSR™ enables the use of culture flasks and other closed vessels, thus facilitating culture scale-up and automation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty