Would you like to log in again?
Human, Mouse, Rat, Non-Human Primate Desoxyribonuclease I (DNase I)
Supplier: Stemcell Technologies
07900
MSPP-07900EA 121.54 USD InStock
MSPP-07900
Human, Mouse, Rat, Non-Human Primate Desoxyribonuclease I (DNase I)
Enzymes
Deoxyribonuclease (DNase) I solution is a cell dissociation reagent useful to reduce or prevent the clumping of concentrated or cryopreserved cell suspensions following thawing.
Contains: Bovine pancreatic deoxyribonuclease I (1 mg/mL) and phosphate-buffered saline
Subtype: Enzymatic
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty