Would you like to log in again?
Siphon Pumps, National Spencer
Supplier: NATIONAL SPENCER, INC. MS
10010 10020
MSPP-10020BX 341.54 USD InStock
MSPP-10020 MSPP-10010
Siphon Pumps, National Spencer
Pumps Drum/Transfer Pumps
Siphon pump made of polyethylene body and polypropylene suction tube.
  • Two years limited warranty
  • Compatible with alcohol, anti-freeze, water-based and windshield washer fluids
Ordering information: Small pump must be ordered in the multiples of 120 and large pump in the multiples of 24.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty