Would you like to log in again?
Sequel Sequencing Kit 3.0 Bundle, Pacific Biosciences
Supplier: Pacific Biosciences
101-642-400
MSPP-101642400EA 4789.23 USD InStock
MSPP-101642400
Sequel Sequencing Kit 3.0 Bundle, Pacific Biosciences
Nucleic Acid Reagents Sanger Sequencing Reagents
Optimized reagents and disposables for performing 20 sequel system SMRT sequencing 3.0 reactions.
The sequel sequencing 3.0 bundle (5 pack - 4 reactions) contains reagent and disposable supplies for 20 reactions of SMRT Sequencing.
Use with sequel SMRT cell 1M v3 and sequel SMRT cell 1M v3 LR and templates prepared with sequel binding and control kit 3.0 to achieve up to 20 Gb for large-insert libraries and more for other applications.
Delivery information: It contains five sequel sequencing kits 3.0 to 4 reaction plate, one sequel SMRT cell oil pack, and one tube septa pack.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty