Would you like to log in again?
Dumont #5CO Forceps, Dumostar, Tip Size 0.05×0.01 mm Biologie Tips, Fine Science Tools
Supplier: Fine Science Tools
11295-20
MSPP-1129520EA 107.69 USD InStock
MSPP-1129520
Dumont #5CO Forceps, Dumostar, Tip Size 0.05×0.01 mm Biologie Tips, Fine Science Tools
Forceps
More durable and corrosion resistant than the best stainless steels.
  • Highly resistant to mineral and organic acids, and salt water
  • Dumostar lasts an average of four times longer than dumoxel and inox
  • Cost effective
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty