Would you like to log in again?
Onyx PCX Derivatization Instruments
Supplier: Pickering Laboratories
Onyx PCX
1155-1099 1155-1071 1155-1081 1155-1105 1155-1103 1155-1101 1155-1061 1155-1011 1155-1031 1155-1021 1155-1096 1155-1051
MSPP-1155-1021EA 40000 USD InStock
MSPP-1155-1021 MSPP-1155-1099 MSPP-1155-1011 MSPP-1155-1101 MSPP-1155-1061 MSPP-1155-1051 MSPP-1155-1096 MSPP-1155-1031 MSPP-1155-1081 MSPP-1155-1071 MSPP-1155-1103 MSPP-1155-1105
Onyx PCX Derivatization Instruments
Chromatography Systems Derivatization Systems
Pickering Laboratories offers the only instrumentation optimized for the analysis of amino acids, carbamates, glyphosate, mycotoxins, antibiotics and many other applications. Each component is specifically designed to enhance sensitivity and selectivity. Only Pickering Laboratories offers complete application support, including chemicals, columns, methods and post-column systems.
  • True pulse-free syringe pump with single piece ceramic barrels
  • Programmable flow rates of 50 to 1500 μl/minute
  • Refill cycle of 60 seconds or less
  • Automatic piston wash, reagent flush cycle
  • Advanced fluidics valve management system
  • Easy access to internal components
For convenient operation and reagent conservation between runs, the Onyx PCX control software operates from a Windows PC. Application conversion from Pinnacle PCX to Onyx PCX is seamless, including method transfers. Temperature gradient column oven with programmable temperature for quicker Amino Acids Analysis run times. Column oven has better tubing protection and is positioned for quick column swapping. Greater sensitivity is provided through an electronic pulse-free syringe pump.
Instrument with doors closed: 56.9×30.5×48.6 cm (22.4 H×12.0 W×19.15" D). Maximum operating pressure 35 bar (500 psi). 27 kg (60 lbs) for systems with two pumps. 23 kg (50 lbs) for single-pump systems. 200 to 240 VAC, 50/60 Hz, 0.8 A, 200 W, or 100 to 120 VAC, 50/60 Hz, 1.7 A, 200 W are the factory default configurations. 10% of normal mains voltage. Maximum current of 5 A. Category II installation (over voltage), degree 2 pollution. up to 1981 m (6500 ft) above sea level. Temperature range: 15 to 29 °C. At 31 °C, the relative humidity can reach 80%. Reactor temperature is heated to a maximum setpoint of 130 °C, which is 5 °C above ambient. 0.5 °C of stability. Precision 1 °C. To prevent, a thermal safety switch restricts temperature to 150 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty