Would you like to log in again?
Phadebas® Amylase Test Kit, Magle Life Science
Supplier: PHADEBAS, INC MS
1301
MSPP-1301EA 244.62 USD InStock
MSPP-1301
Phadebas® Amylase Test Kit, Magle Life Science
Assays Enzyme Assays
Phadebas® Amylase test tablets are used for accurately determining α-amylase activity for a wide variety of applications, from different fields in the food industry to biotech and detergents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty