Would you like to log in again?
EasySep™ Human T Cell Isolation Kit, Stemcell Technologies
Supplier: Stemcell Technologies
17951
MSPP-17951EA 1483.08 USD InStock
MSPP-17951
EasySep™ Human T Cell Isolation Kit, Stemcell Technologies
Assays Cellular Assays
The EasySep™ human T cell isolation kit is designed to isolate T cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection.
  • Fast, easy-to-use and column-free
  • Up to 98% purity with high recovery
  • Untouched, viable cells
  • 8-Minute cell isolation kit using immunomagnetic negative selection
  • Sample source: Leukapheresis, PBMC
  • Selection method: Negative
The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
Magnet compatibility includes EasySep™ magnet, the big easy EasySep™ magnet, easy 50 EasySep™ magnet, EasyPlate™ EasySep™ magnet, EasyEights™ EasySep™ magnet and RoboSep™-S.
Delivery information: Components include EasySep™ human T cell isolation kit, EasySep™ human T cell isolation cocktail (1 ml), EasySep™ dextran RapidSpheres™ (1 ml), RoboSep™ human T cell isolation kit, EasySep™ human T cell isolation cocktail (1 ml), EasySep™ dextran RapidSpheres™ (1 ml), RoboSep™ buffer and RoboSep™ filter tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty