Would you like to log in again?
Instant Ocean Aquarium Salt, That Fish Place
Supplier: THAT FISH PLACE MS
198262
MSPP-198262EA 83.06 USD InStock
MSPP-198262
Instant Ocean Aquarium Salt, That Fish Place
Specimens Live Specimens Aquaria - Living Material
Instant ocean synthetic sea salt contains every necessary major, minor, and trace element and has no nitrates and no phosphates.
  • Developed through sophisticated biological and chemical testing
  • Every batch is analyzed to assure consistent high quality
  • It comes with exceptional solubility, uniform particle size
Mix with ordinary dechlorinated tap or purified water. To prepare small quantities, use half a cup of mix per gallon of water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty