Would you like to log in again?
EasySep™ Mouse CD8+ T Cell Isolation Kit, Stemcell Technologies
Supplier: Stemcell Technologies
19853
MSPP-19853EA 1216.92 USD InStock
MSPP-19853
EasySep™ Mouse CD8+ T Cell Isolation Kit, Stemcell Technologies
Assays Cellular Assays
Isolate untouched and highly purified CD8+ T cells from mouse splenocytes by immunomagnetic negative selection. When using single-cell suspensions from
other tissue types, this kit may require optimization.
  • Fast and easy-to-use
  • Up to 95% purity
  • No column required
  • Untouched, viable cells
This kit targets non-CD8+ T cells for removal with biotinylated antibodies recognizing specific cell surface markers. Unwanted cells are labeled with biotinylated
antibodies and streptavidin-coated magnetic particles, and separated without columns using an EasySep™ magnet. Desired cells are simply poured off into a
new tube. Isolated cells are immediately available for downstream applications such as flow cytometry, culture, and cell-based experiments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty