Would you like to log in again?
Ear Punch, Fine Science Tools
Supplier: Fine Science Tools
24214-02
MSPP-2421402EA 124.62 USD InStock
MSPP-2421402
Ear Punch, Fine Science Tools
Biopsy Punches
This ear punch is designed to ensure a fast and clean punch.
  • Light weight
  • Fits comfortably in the palm
  • Autoclavable
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty