Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tissue-Tek® Cryomolds® Molds, Ted Pella Inc

Supplier: Ted Pella

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Tissue-Tek® Cryomold®
27183 27180
MSPP-27180PK 53.31 USD InStock
MSPP-27180 MSPP-27183
Tissue-Tek® Cryomolds® Molds, Ted Pella Inc
Moulds Moulds for Histology
For use with Tissue-Tek® II M/C.

Tissue-Tek Cryomold® creates a uniformly shaped, flat-surface specimen blocks with O.C.T. Compound for frozen sectioning.

Disposable PVC plastic molds fit Cryobar® freezing wells.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty