Would you like to log in again?
Blunt-End Needles, 16 Gauge, Stemcell Technologies
Supplier: Stemcell Technologies
28110
MSPP-28110PK 318.46 USD InStock
MSPP-28110
Blunt-End Needles, 16 Gauge, Stemcell Technologies
Syringe Needles
Ideal for safe and accurate handling of methylcellulose-based media. Blunt end reduces the risk of needle stick injuries.
  • With a metal Luer lock hub
Manufactured from non-pyrogenic, non-toxic materials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty