Would you like to log in again?
Trypsin
Supplier: 0000013421
30-2101
MSPP-30-2101EA 26.15 USD InStock
MSPP-30-2101
Trypsin
Enzymes
This solution contains 0.25% Trypsin/0.53 mM EDTA in Hanks balanced salt solution without calcium or magnesium. For dissociation of cell monolayers.
Trypsin-EDTA solution is suitable for most but not for all adherent cell lines. This product contains phenol red.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty