Would you like to log in again?
AquaLab Cleaning Kit, Meter Group
Supplier: Meter Group
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
30595
MSPP-30595EA 161.54 USD InStock
MSPP-30595
AquaLab Cleaning Kit, Meter Group
Wipes Cleaning Kits and Pads
This kit is used to clean benchtop water activity meter, a portable water activity meter, or a vapor sorption analyzer.
Delivery information: The kit includes distilled water for rinsing, small kimwipes, large kimwipes, plastic rods, charcoal, and a DVD with detailed instructions.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty