Would you like to log in again?
Hepatitis B Surface Antigen (HBsAg, native or recombinant) ELISA Kit, Alpha Diagnostic
Supplier: ALPHA DIAGNOSTIC INTERNAT MS
4110
MSPP-4110EA 830.77 USD InStock
MSPP-4110
Hepatitis B Surface Antigen (HBsAg, native or recombinant) ELISA Kit, Alpha Diagnostic
Assays ELISAs
Quantitative ELISA Kit is used for the measurement of recombinant HBsAg in manufacturing process or vaccine formulations.
Caution: For research use only, not for diagnostic or therapeutic use.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty