Would you like to log in again?
GripTips for Electronic Pipetting Systems, INTEGRA Biosciences AG
Supplier: Integra Biosciences
GripTips
4403 4405 4455 4454 4446 4414 4433 4436 4432 4435 4412 4404 4426 4443 4422 4444 4425 4434 4423 4424 4445
MSPP-4405PK 333 USD InStock
MSPP-4405 MSPP-4404 MSPP-4426 MSPP-4403 MSPP-4414 MSPP-4436 MSPP-4425 MSPP-4446 MSPP-4435 MSPP-4424 MSPP-4412 MSPP-4434 MSPP-4423 MSPP-4445 MSPP-4455 MSPP-4433 MSPP-4422 MSPP-4444 MSPP-4454 MSPP-4432 MSPP-4443
GripTips for Electronic Pipetting Systems, INTEGRA Biosciences AG
Pipette Tips
These GripTips are manufactured from 100% virgin polypropylene under strictly controlled conditions. Without hammering, they snap onto the pipette's tip fitting, which prevents tips from loosening, leaking or falling off.
  • Low retention
  • Extended length design
  • Tips can be autoclaved
  • Manufactured from 100% virgin polypropylene under strictly controlled conditions
  • These are free of RNase, DNase, pyrogens, PCR inhibitors and endotoxins
The rim of the GripTips will snap over the multi-lobes, attaching firmly. As a result, they will never accidently fall off. A shoulder provides a positive stop to prevent over-tightening of the tip. It can only be attached up to that point and no further, ensuring that all tips are at the same height. As a result, the tip is either on or off, but nothing in between.
Viscous samples, detergents and other low surface tension liquids tend to interact with standard pipette tips forming a thin liquid film on the pipette tip inner wall. This effect leads to pipetting inaccuracy and inconsistency as well as loss of precious reagents. A unique polypropylene blend is used to mold low retention GripTips with heightened hydrophobic properties. Using low retention these tips prevents low surface tension samples from spreading out and 'wetting' the inner wall of the tips, allowing them to bead-up for a maximum liquid recovery.
They feature an extended length design enabling easy access into labware types such as tubes and deep well blocks. This extended length design also reduces PCR cross contamination as the elongated tip prevents the pipette shaft from contacting the wall of the PCR tubes.
An environmentally friendly refill system allows the reuse of the current tip racks and thus reduces plastic waste by 60%. Tip racks with single inserts in 96 and 384 fold configuration. The 384 fold configuration is highly space efficient and allows tip loading with a 16-channel pipette.
Packaging: The economical bulk packs option.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty