Would you like to log in again?
VOYAGER II Adjustable Tip Spacing Pipetting Systems, INTEGRA Biosciences AG
Supplier: Integra Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VOYAGER
4722 4723
MSPP-4723EA 3744.62 USD InStock
MSPP-4723 MSPP-4722
VOYAGER II Adjustable Tip Spacing Pipetting Systems, INTEGRA Biosciences AG
Pipettes Repetitive Pipettors
VOYAGER II pipettes feature with motorized tip spacing which helps in enabling parallel transfer of multiple samples between labware of different sizes and formats.
  • GripTips snap into place with minimal tip loading effort, providing a secure connection
  • Pipettes can be charged directly with the pipette mains adapter
  • Versatile user interface and ergonomic design
  • Dispensing multiple aliquots of the same volume
VOYAGER II pipettes feature with motorized tip spacing which helps in enabling parallel transfer of multiple samples between labware of different sizes and formats.
These pipettes provide access to a wide range of labware formats such as microplates, tube racks or gel chambers. These are ideal for a wide variety of genomic, proteomic and cell culture applications.
Ordering information: Power supplies are not included, need to order separately.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty