Would you like to log in again?
deconex® 11 UNIVERSAL, Cleaning Concentrate for Laboratory Glassware, TechnoTrade International
Supplier: TECHNO TRADE INTER MS
deconex®
500100.00-K05W
MSPP-50010000KEA 211.54 USD InStock
MSPP-50010000K
deconex® 11 UNIVERSAL, Cleaning Concentrate for Laboratory Glassware, TechnoTrade International
Cleaning Agents Detergents and Surface Cleansers
deconex® 11 UNIVERSAL is a mildly alkaline, liquid concentrate with activated chlorine used for the cleaning and decontamination of laboratory glassware.
deconex® 11 UNIVERSAL exhibits a wide cleaning spectrum due to its composition. This allows even stubborn contaminants to be removed such as baked-on fats, oils, waxes, dried blood, proteins, residues of culture media, pen markings, pigments, traces of organic and inorganic materials and silicones.
Suitable for: Borosilicate glass 3.3, Duran®, quartz glass, porcelain, stainless steel, chrome nickel steel, polyethylene, polypropylene, Teflon®, silicone rubber, natural rubber.
Not suitable for: Aluminium, zinc, tin
For materials not mentioned please make your own specific compatibility tests or consult Borer Chemie AG
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty