Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Evaporation oil, ibidi

Supplier: Ibidi
50051
MSPP-50051EA 492.31 USD InStock
MSPP-50051
Anti-Evaporation oil, ibidi
Stains Microscopy and staining reagents
A biocompatible silicone oil used to prevent medium evaporation in cell culture applications.

  • Excellent reduction of medium evaporation
  • Ready to use
  • Dry-heat sterilized for direct use
  • Viscosity optimized for cell culture
  • Compatible with microscopy
  • Compatible with all ibidi products and cell culture applications

Specifications
Appearance: Optically clear in dry atmosphere
Density (20°C): 1.05 g/ml
Viscosity (20°C): Ca. 200 mPa ∙ s
Refractive index nD: 1.453
Temperature stability: 121°C (dry-heat only)
Storage of product: Dark, room temperature

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty