Would you like to log in again?
Dumont Anti-Capillary Tweezers, Ted Pella
Supplier: Ted Pella
510-4NM
MSPP-510-4NMEA 88.34 USD InStock
MSPP-510-4NM
Dumont Anti-Capillary Tweezers, Ted Pella
Forceps
This anti-capillary, reverse (self closing) and regular tweezers are great for handling transmission electron microscopy grids. The picture above shows the anti-capillary tweezer holding an electron microscopy grid. The tweezers tips are shaped and aligned to pick up less moisture when removing grids from a liquid.
  • Biological Grade
  • The tips are delicate and can become misaligned
  • Gentle finger pressure on the blades can be used to realign the tips
The top (bent) tip meets the bottom(flat) tip at a slight distance behind the point, thus forming a small lip which supports the grid and allows it to be picked up without twisting. There is no guarantee against breakage of the tips for any of the anti-capillary style tweezers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty