Would you like to log in again?
tissueTUBE Sample Tubes with Plugs, Covaris®
Supplier: Covaris
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
520001 520006 520036
MSPP-520006CS 141.54 USD InStock
MSPP-520006 MSPP-520036 MSPP-520001
tissueTUBE Sample Tubes with Plugs, Covaris®
Tubes Test Tubes
These tubes are suitable for sample collection, storage, pulverization, and transfer.
  • Ideal for samples of mass 1 g
Used with the CP02 cryoprep pulverizer and with holder tissueTUBE TT1 and tissueTUBE extra thick.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty