Would you like to log in again?
Microcystins-ADDA ELISA (Microtiter Plate) Kit, Abraxis
Supplier: GOLD STD DIAGNOSTICS HORSHAM MS
520011
MSPP-520011PK 853.92 USD InStock
MSPP-520011
Microcystins-ADDA ELISA (Microtiter Plate) Kit, Abraxis
Assays ELISAs
The Microcystins-ADDA ELISA is an enzyme-linked immunosorbent assay for the quantitative and sensitive congener-independent detection of microcystins and nodularins in water samples.
No additional sample preparation is required prior to analysis. Positive samples can be confirmed by HPLC, protein phosphatase assay, or other conventional methods.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty