Would you like to log in again?
Dispase (from Bacillus polymyxa), Stemcell Technologies
Supplier: Stemcell Technologies
07913
MSPP-7913EA 443.08 USD InStock
MSPP-7913
Dispase (from Bacillus polymyxa), Stemcell Technologies
Enzymes
Dispase is a protease that is suitable for the gentle dissociation of a wide variety of tissues.
It contains 5 U/ml dispase II (neutral protease from Bacillus polymyxa) dissolved in Hanks’ balanced salt solution.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty