Would you like to log in again?
µ-Slide Microscopy Rack, Ibidi
Supplier: IBIDI USA MS
80030
MSPP-80030EA 546.15 USD InStock
MSPP-80030
µ-Slide Microscopy Rack, Ibidi
Microscope Slide Boxes
A microscopy rack for perfect µ-Slide stabilization.
  • Multiwell format is compatible with all inverted microscopes
  • Lid can be fixed with magnetic positioning for additional stabilization
  • Magnetic lids not included, must be ordered separately
The µ-Slide microscopy rack is an aluminum rack made to hold 4 µ-Slides for analysis of multiple samples. The multiwell format is compatible with all inverted microscopes. Rack may be sterilized via autoclave or alcohol. Magnetic lids are sold seperately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty