Would you like to log in again?
Clamp for sticky-Slides, Ibidi
Supplier: IBIDI USA MS
80040
MSPP-80040EA 176.92 USD InStock
MSPP-80040
Clamp for sticky-Slides, Ibidi
Clamps General Purpose Clamps
This instrument securely fastens sticky-Slides to coverslip bottoms.
  • Applies homogeneous force for tight sealing, with no leakage
  • Clamping force: 1.2 kN
  • Easy to use for an optimal bond; clamp must be paired with one adapter
  • Clamp and adapters must be ordered separately
The clamp creates optimal adhesive bond between sticky-Slide and coverslip to provide a tight seal.
Ordering information: Adapters sold seperately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty