Would you like to log in again?
sticky-Slide I 0.4 Luer Slide, Ibidi
Supplier: IBIDI USA MS
80178
MSPP-80178PK 584.62 USD InStock
MSPP-80178
sticky-Slide I 0.4 Luer Slide, Ibidi
Slides Chamber Slides
A bottomless channel slide used for perfusion applications with a self-adhesive underside to which own substrates can be mounted. 0.4 mm channel height.
  • Versatile slide permits the mounting of a variety of bottom materials
  • Channel length: 50 mm
  • Channel height: 400+50 µm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty