Would you like to log in again?
Quantum™ Simply Cellular® Microspheres, Bangs Laboratories
Supplier: Bangs Laboratories
816B 815A 816 815B 817 817A 816A
MSPP-816EA 1283.08 USD InStock
MSPP-816 MSPP-817 MSPP-816A MSPP-817A MSPP-815B MSPP-816B MSPP-815A
Quantum™ Simply Cellular® Microspheres, Bangs Laboratories
Beads Microcarriers
Quantum™ Simply Cellular® (QSC) microspheres are used in the quantitative analysis of cellular antigen expression. When stained with the same antibody that is used to label cells, they permit determination of the Antibody Binding Capacity (ABC) of the cells.
  • QSC kit consists of 5 bead populations - 1 blank and 4 with increasing levels of Fc-specific capture antibody
These kits are comprised of five microsphere populations that are 7 to 9 µm in diameter: one blank and four labeled with increasing amounts of antibody. Each coated population binds a specific number of monoclonal antibodies of the noted species, which is equal to its Antibody Binding Capacity (ABC) value. The investigator labels QSC beads using the same antibodies used to stain cells. The free QuickCal® analysis template aids in determining the expression level of cells, and for evaluating instrument linearity and detection threshold.
The coated populations are calibrated in terms of their ABC for monoclonal antibodies, and beads are labeled to saturation with the same monoclonal that is used to label cells. Beads are run in the same type of suspending solution/medium, on the same instrument, at the same instrument settings (PMT voltages, compensation settings), and on the same day as cell samples.
Ordering information: Quantum™ Simply Cellular® kits are available in two sizes: A (20 tests), B (100 tests). A free QuickCal® analysis template is provided with each kit.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty