Would you like to log in again?
Mouse Leptin ELISA Kit, Crystal Chem
Supplier: CRYSTAL CHEM INC MS
90030
MSPP-90030EA 684.62 USD InStock
MSPP-90030
Mouse Leptin ELISA Kit, Crystal Chem
Assays ELISAs
The leptin mouse ELISA is used to quantify levels of mouse leptin in serum, plasma, cell culture, and fluid using only a 5 µl sample volume.
  • High sensitivity
  • Designed for mouse samples
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty