Would you like to log in again?
Ultra Sensitive Mouse Insulin ELISA Kit, Crystal Chem
Supplier: CRYSTAL CHEM INC MS
90080
MSPP-90080EA 653.85 USD InStock
MSPP-90080
Ultra Sensitive Mouse Insulin ELISA Kit, Crystal Chem
Assays ELISAs
The insulin mouse Ultra sensitive ELISA is used to quantify levels of mouse insulin in serum, plasma, cell culture, and fluid using only a 5 µl sample volume.
  • Kits use only 5 µl sample
  • Dynamic range of 0.1 to 64 ng/ml
  • High sensitivity (50 pg/ml) using 5 µl sample
  • Specific and fast
  • Recovery: When mouse insulin was spiked to mouse serum, the recovery of mouse insulin was 100% ±15%
Ultra sensitive mouse insulin ELISA kit have the low range assay of 0.1 to 6.4 ng/ml, wide range assay of 0.1 to 12.8 ng/ml and high range assay of 1 to 64 ng/ml.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty