Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SmartEnzymes™ FabRICATOR® Z Fab2 Kit Microspin, Genovis

Supplier: Genovis
A2-FZ2-025 A2-FZ2-005
MSPP-A2FZ2025EA 2930.77 USD InStock
MSPP-A2FZ2025 MSPP-A2-FZ2005
SmartEnzymes™ FabRICATOR® Z Fab2 Kit Microspin, Genovis
Protein Purification Protein Purification Kits
FabRICATOR Z Fab2 Kit consists of spin columns of FabRICATOR Z Immobilized for antibody digestion, and spin columns of CaptureSelect™* Fc resin for purification of fragments. One FabRICATOR Z spin column contains sufficient FabRICATOR Z-coupled agarose beads to process 0.5 mg mouse IgG2a and IgG3.

  • Purification of homogenous F(ab')2 fragments from mouse IgG2a and IgG3
  • Fast - pure fragments in under two hours
  • Reliable workflow for preparative applications

Product Composition: FabRICATOR Z Immobilized microspin column containing 0.1 ml settled resin of FabRICATOR Z enzyme immobilized on agarose. CaptureSelect Fc microspn column containing 0.1 ml settled resin of CaptureSelect™ Fc₁ affinity ligand immobilized on agarose. Agarose resins are suspended in 20% ethanol/water (v/v).

The immobilized enzyme and affinity resin helps for hinge digestion of mouse IgG2a and IgG3 and purification of fragments.

One spin column contains sufficient FabRICATOR Z-coupled agarose beads to digest 0.5 mg mouse IgG2a or IgG3.

The FabRICATOR Z Immobilized spin columns and CaptureSelect™* spin columns are supplied in 20% ethanol, shipped cold, and should be stored at 4 to 8 °C upon arrival. Do not freeze the products.

Appearance: White suspension.
Binding capacity: >15 mg hIgG per ml CaptureSelect Fc resin.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty