Would you like to log in again?
Universal Vials, Ab Sciex
Supplier: AB Sciex
A62251
MSPP-A62251EA 131.63 USD InStock
MSPP-A62251
Universal Vials, Ab Sciex
Vials
Universal vials are used to accommodate buffers/samples.
These polymethylpentene vials are precision manufactured and have been compatibility tested with commonly used CE reagents. Universal vials are designed for use only with CESI 8000, PA 800 Enhanced or PA 800 plus instruments and should not be reused.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty