Would you like to log in again?
HEPA Filter, Airclean Systems
Supplier: Airclean Systems
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ACFHEPA
MSPP-ACFHEPAEA 538.46 USD InStock
MSPP-ACFHEPA
HEPA Filter, Airclean Systems
Filters Gas Filters
High Efficiency Particulate Air (HEPA) Filters are available for the containment of particles.
The filter has a 99.997% efficiency rating for collecting 0.3 µm particles.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty