Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Normocin™ - Antimicrobial Reagent, InvivoGen

Supplier: InvivoGen
ANT-NR-2 ANT-NR-1
MSPP-ANTNR1EA 286.15 USD InStock
MSPP-ANTNR1 MSPP-ANTNR2
Normocin™ - Antimicrobial Reagent, InvivoGen
Media for Microbiology Additives and Supplements for Microbiology
Normocin™ is an innovative antibiotic formulation designed to prevent cell lines from mycoplasma, bacterial and fungal contaminations. Normocin™ can be routinely added in the cell culture medium in combination with penicillin/streptomycin to broaden the anti-bacterial spectrum.

  • Active against both gram+ and gram- bacteria, mycoplasmas and fungi
  • Can be used in combination with penicillin/streptomycin
  • Non-toxic to the cell line being treated
  • One 1 ml vial of Normocin™ is sufficient to treat 500 ml of culture
  • One 20 ml bottle of Normocin™ is sufficient to treat 10 liters of culture
  • Active concentration not toxic for treated cell lines: 100 μg/ml
  • Endotoxin level: <0.5 EU/mg

Normocin™ contains three compounds. Two of these compounds act on gram+ bacteria, gram-acteria, and mycoplasmas by blocking DNA and protein synthesis. The third compound eradicates yeasts and fungi by disrupting ionic exchange through the cell membrane.

Normocin™ should be stored at 4 °C or –20°C.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty