Would you like to log in again?
Accutase® Cell Detachment Solution
Supplier: Innovative Cell Technologies
AT 104 AT104-500
MSPP-AT104EA 41.54 USD InStock
MSPP-AT104 MSPP-AT104500
Accutase® Cell Detachment Solution
Enzymes
Accutase® is a ready to use non-mammalian, non-bacterial replacement for all applications of trypsin.
  • Can be used whenever gentle and efficient detachment of any adherent cell line is needed
  • Works extremely well on embryonic and neuronal stem cells; mono layers of stem cells can be grown after passaging with Accutase
  • Preserves most epitopes for subsequent flow cytometry analysis
  • Does not need to be aliquoted
  • Bottle is stable in the refrigerator for 2 months
It is a natural enzyme mixture with proteolytic and collagenolytic enzyme activity. This means it mimics the action of trypsin and collagenase at the same time. However, because it is more efficient than mammalian trypsin and collagenase, it is formulated at a much lower concentration making it less toxic and gentler, but just as effective.
Accutase® does not need to be neutralized when passaging adherent cells. The addition of more media after the cells are split dilutes Accutase so it is no longer able to detach cells.
Applications include hESC culturing, analysis of cell surface markers, virus growth assay, quiescence assays by serum starvation, transformation assays by oncogene transfection, neural crest cell migration assays, cell proliferation, apoptosis, cell haptotaxsis, tumor cell migration assays, routine cell passage, production scale-up (bioreactor), and flow cytometry.
hESCs, fibroblasts, keratinocytes, vascular endothelial cells, hepatocytes, vascular smooth muscle cells, hepatocyte progenitors, primary chick embryo neuronal cells, bone marrow stem cells, adherent CHO and BHK cells, macrophages, 293 cells, L929 cells, immortalized mouse testicular germ cells, 3T3, Vero, COS, HeLa, NT2, MG63, M24 and A375 metastatic melanoma, gliomas U251 and D54, HT1080 fibrosarcoma cells, and Sf9 insect cells can be detach by Accutase without harm.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty