Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lympholyte®-M Cell Separation Media, Cedarlane

Supplier: Cedarline
CL5031 CL5030
MSPP-CL5030EA 284.62 USD InStock
MSPP-CL5030 MSPP-CL5031100
Lympholyte®-M Cell Separation Media, Cedarlane
Cell Separation Media
Lympholyte®-M is a density separation medium specifically designed for the isolation of viable lymphocytes from murine lymphoid cell suspensions.

  • Composition: Polysucrose 400 and sodium diatrizoate
  • Density: 1.0875 ±0.0010 g/cm3 at 22 °C
  • pH: 6.9 ±0.3
  • Viability/Purity: Recovery of viable lymphocytes >70%

Lympholyte®-M can be utilized in a simple protocol to eliminate erythrocytes, dead cells and debris from murine spleen, lymph node, thymus and bone marrow suspensions. The resulting cell population demonstrates a high and nonselective recovery of viable lymphocytes that are suitable for use as target cells in cytotoxicity, FACS assays, and in in vivo and in vitro functional studies.

Applications include the removal of dead cells in sequential cytotoxicity studies, the removal of erythrocytes, dead cells and debris from other murine tissue
suspensions including liver and lung, the harvesting of viable cells and removal of dead cells and debris from various clone cell and hybridoma cell lines and the isolation of murine nuclear epidermal cells.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty